missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98421B
This item is not returnable.
View return policy
Description
UCH-L3 Polyclonal specifically detects UCH-L3 in Rat samples. It is validated for Western Blot, ELISA.
Specifications
| UCH-L3 | |
| Polyclonal | |
| Western Blot, ELISA | |
| EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3 | |
| Produced in rabbits immunized with purified, recombinant UCH-L3 (Q91Y78; Glu 2-Ala 230). | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| Western Blot, ELISA | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7347 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction