Learn More
UCHL1/PGP9.5 Mouse anti-Human, CoraLite 488, Clone: 1C9E11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66230-100UL
Product Code. 16820756
  • € 450.00 / 100µL
Description
The UCHL1/PGP9.5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UCHL1/PGP9.5. This antibody recognizes human, mouse, pig, rat antigen. The UCHL1/PGP9.5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
UCHL1/PGP9.5
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1C9E11
CoraLite 488
100 μL
7345