Learn More
UCHL1/PGP9.5 Mouse anti-Human, CoraLite 488, Clone: 1C9E11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66230-100UL
Description
The UCHL1/PGP9.5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UCHL1/PGP9.5. This antibody recognizes human, mouse, pig, rat antigen. The UCHL1/PGP9.5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
| UCHL1/PGP9.5 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1C9E11 | |
| CoraLite 488 | |
| 100 μL | |
| 7345 | |
| IgG1 |