missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCK1 Monoclonal Antibody (OTI6H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501490
This item is not returnable.
View return policy
Description
Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4- thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)- benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5- methylcytidine, and N(4)-anisoylcytidine.Specifications
| UCK1 | |
| Monoclonal | |
| 0.72 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HA47 | |
| UCK1 | |
| Full length human recombit protein of human UCK1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI6H7 | |
| Unconjugated | |
| UCK1 | |
| URK1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 83549 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction