Learn More
UCK1 Mouse anti-Human, Clone: OTI4C2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501473
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4- thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)- benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5- methylcytidine, and N(4)-anisoylcytidine.Specifications
| UCK1 | |
| Monoclonal | |
| Unconjugated | |
| UCK1 | |
| URK1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 83549 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4C2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9HA47 | |
| UCK1 | |
| Full length human recombit protein of human UCK1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.