Learn More
Invitrogen™ UCKL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590762
Description
Immunogen sequence: MAAPPARADA DPSPTSPPTA RDTPGRQAEK SETACEDRSN AESLDRLLPP VGTGRSPRKR TTSQCKSEPP LLRTSKRTIY TAGRPPWYNE HGTQSKEAFA IGLGGGSASG KTTVARMIIE ALDVPWVVLL SMDSFYKVLT EQQQEQAAHN NFNFDHPDAF DFDLIISTLK KLKQGKSVKV PIYDFTTHSR KKDWKTLYGA; Positive Samples: Mouse heart, Mouse lung, Rat pancreas; Cellular Location: Cytoplasm, Nucleus.
Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
Specifications
| UCKL1 | |
| Polyclonal | |
| Unconjugated | |
| Uckl1 | |
| 1110007H10Rik; F538; RGD1565465; UCK1L; UCK1-LIKE; Uckl1; uridine kinase-like 1; uridine-cytidine kinase 1 like 1; uridine-cytidine kinase 1-like 1; uridine-cytidine kinase-like 1; URKL1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 499956, 54963, 68556 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.29 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91YL3, Q9NWZ5 | |
| Uckl1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human UCKL1 (NP_0603292). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.