missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCN3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142332
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Urocortin 3 (UCN3) is a member of the corticotropin-releasing factor (CRF) family, encoded by the UCN3 gene, and is primarily involved in stress response and energy balance regulation. UCN3 is predominantly expressed in the hypothalamus and certain peripheral tissues, such as pancreatic islets, where it plays significant roles in modulating anxiety-related behaviors and metabolism. This neuropeptide functions by binding to CRF2 receptors, exerting effects on glucose metabolism and insulin secretion. UCN3 has been implicated in protecting the heart from ischemic damage and modulating the cardiovascular system's response to stress, highlighting its potential in treating cardiac disorders. Additionally, it influences nutrient partitioning and energy homeostasis, with expression levels affected by nutritional status. Research indicates that UCN3 deficiency may lead to impaired glucose tolerance and altered energy expenditure, demonstrating its importance in metabolic health. Furthermore, UCN3 has been associated with the regulation of neuroendocrine signaling pathways, providing insights into therapeutic strategies for metabolic and stress-related disorders.
Specifications
| UCN3 | |
| Polyclonal | |
| Unconjugated | |
| UCN3 | |
| prepro-urocortin 3; RGD1562213; SCP; SPC; Stresscopin; ucn III; Ucn3; UCNIII; urocortin 3; urocortin 3 (stresscopin); Urocortin II; Urocortin III; Urocortin-3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 114131 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q969E3 | |
| UCN3 | |
| Peptide with sequence C-KPRQDTAKSPHRTK (aa 106-119). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction