missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP1/3 Antibody (SR1570), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | UCP1/3 |
|---|---|
| Clone | SR1570 |
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659244
|
Novus Biologicals
NBP3-22657-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619144
|
Novus Biologicals
NBP3-22657-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCP1/3 Monoclonal antibody specifically detects UCP1/3 in Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| UCP1/3 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR1570 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| A synthesized peptide derived from human UCP1/3 (Uniprot #: P12242, P56501) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title