missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-2828-25ug
This item is not returnable.
View return policy
Description
UCP1 Polyclonal specifically detects UCP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.
Specifications
| UCP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Rat and Dog. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1 μg/mL, Peptide ELISA Detection limit 1:128000 | |
| NP_068605.1 | |
| UCP1 | |
| Peptide with sequence C-EQLKRELSKSRQ corresponding to C-Terminus according to NP_068605.1. | |
| 25 μg | |
| Cancer, Cellular Signaling, metabolism | |
| 7350 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction