Learn More
UCP2 Antibody (3F1B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52582
 View more versions of this product
Product Code. 18167899
  • € 552.00 / 0.10mg
Description
UCP2 Monoclonal specifically detects UCP2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UCP2
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready
mitochondrial uncoupling protein 2, SLC25A8BMIQ4, Solute carrier family 25 member 8, UCP 2, UCPH, uncoupling protein 2 (mitochondrial, proton carrier)
Mouse
33.2 kDa
0.1 mg
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
3F1B9
Unconjugated
PBS with 0.05% Sodium Azide
UCP2
Purified recombinant fragment of human UCP2 (AA: 1-309) expressed in E. Coli.
Protein A or G purified
RUO
7351
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2 α