missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 593.00
Specifications
| Antigen | UCP3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225372
|
Novus Biologicals
NBP2-58840 |
100 μL |
€ 593.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623348
|
Novus Biologicals
NBP2-58840-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCP3 Polyclonal specifically detects UCP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| UCP3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 7352 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AEVVTYDILKEKLLDYHLLTDNFPCHFV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| mitochondrial uncoupling protein 3, SLC25A9Solute carrier family 25 member 9, UCP 3, uncoupling protein 3 (mitochondrial, proton carrier), Uncoupling protein-3 | |
| UCP3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title