missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UCP3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17727597
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17727597 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17727597 Supplier Invitrogen™ Supplier No. PA1055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-055 detects uncoupling protein-3 (UCP-3) mouse, rat, and canine tissues. This antibody will also detect recombinant human UCP-3, but does not detect UCP-3 from mouse liver or kidney. PA1-055 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~30 kDa protein representing UCP-3 from rat heart homogenate. The PA1-055 immunogen is a synthetic peptide corresponding to residues C R(295) A L M K V Q V L R E S P F(308) of mouse and rat UPC-3. This peptide differs from the porcine and human UCP-3 sequence by a single amino acid substitution. PA1-055 immunizing peptide (Cat. # PEP-074) is available for use in neutralization and control experiments.

The uncoupling protein UCP1 (formerly designated UCP) is an integral membrane protein unique to brown adipose tissue mitochondria. UCP1 forms a dimer that acts as a proton channel, which can uncouple oxidative phosphorylation by dissipating the electrochemical potential across the inner mitochondrial membrane. This process induces heat production in brown adipose tissue and is involved in regulation of body temperature and glucose metabolism. UCP2 is a structurally related protein that also uncouples mitochondrial respiration. It is more widely expressed in human and mouse tissues, including white adipose tissue and muscle, than is UCP1. UCP2 is thought to play a role in body weight regulation. An additional UCP family member, UCP3, is highly muscle specific and is possibly involved in the uncoupling of oxidative phosphorylation in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene Ucp3
Gene Accession No. P56499, P56501, Q9N2I9
Gene Alias AI645527; mitochondrial uncoupling protein 3; Slc25a9; solute carrier family 25 member 9; UCP 3; UCP3; UCP-3; uncoupling protein; uncoupling protein 3; uncoupling protein 3 (mitochondrial, proton carrier); uncoupling protein 3, mitochondrial
Gene Symbols Ucp3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C R(295) A L M K V Q V L R E S P F(308) of mouse and rat UCP-3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22229, 25708, 403575
Target Species Mouse, Rat, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.