missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534952
This item is not returnable.
View return policy
Description
Recommended positive controls: SK-N-SH, IMR32, SK-NAS. Predicted reactivity: Mouse (95%), Rat (96%), Dog (93%), Pig (90%), Sheep (93%), Rhesus Monkey (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UCP4 is a 323 amino acid mitochondrial uncoupling protein belonging to the large mitochondrial substrate carrier family with three Solcar repeats. UCP4 is a mitochondrial transport carrier that facilitates the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. It reduces the mitochondrial membrane potential in mammalian cells and is involved in thermoregulatory heat production. UCP4 creates proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation from ATP synthesis resulting in dissipation of energy in the form of heat. UCP4 is usually found in adult and fetal brain, with low levels in spinal chord, corpus callosum and substantia nigra.
Specifications
| UCP4 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A27 | |
| 3632410G24Rik; 9430092A03Rik; D530043E16Rik; Mitochondrial uncoupling protein 4; slc25a27; Solute carrier family 25 member 27; solute carrier family 25, member 27; UCP 4; ucp4; uncoupling protein; uncoupling protein 4; uncoupling protein UCP-4; UNQ772/PRO1566; zgc:63899 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95847 | |
| SLC25A27 | |
| Recombinant protein encompassing a sequence within the center region of human UCP4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction