missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94631-0.02ml
This item is not returnable.
View return policy
Description
UCP5 Polyclonal antibody specifically detects UCP5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UCP5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BMCP-1, BMCP1UCP 5, brain mitochondrial carrier protein 1, Mitochondrial uncoupling protein 5, solute carrier family 25 (mitochondrial carrier, brain), member 14, Solute carrier family 25 member 14, UCP5MGC149543 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-200 of human UCP5 (NP_001269124.1). GHQKSTTVSHEMSGLNWKPFVYGGLASIVAEFGTFPVDLTKTRLQVQGQSIDARFKEIKYRGMFHALFRICKEEGVLALYSGIAPALLRQASYGTIKIGIYQSLKRLFVERLEDETLLINMICGVVSGVISSTIANPTDVLKIRMQAQGSLFQGSMIGSFIDIYQQEGTRGLWRGV | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9016 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction