Learn More
UEVLD Monoclonal Antibody (OTI11E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809207S
This item is not returnable.
View return policy
Description
UEVLD Monoclonal antibody specifically detects UEVLD in Human samples. It is validated for Western BlotSpecifications
| UEVLD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IX04 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55293 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI11E2 | |
| Unconjugated | |
| UEVLD | |
| 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD | |
| Human recombinant protein fragment corresponding to amino acids 131-379 of human UEVLD produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |