missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFD1L Antibody (OTI8G6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74781IR
This item is not returnable.
View return policy
Description
UFD1L Monoclonal specifically detects UFD1L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UFD1L | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UFD1L | |
| Full length recombinant protein of human UFD1L (NP_005650) produced in E.coli. | |
| 0.1 mL | |
| 7353.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI8G6 | |
| Western Blot, Immunohistochemistry | |
| ATP1C, ATP1G1ATPase, Na+/K+ transporting, gamma 1 polypeptide, FXYD domain containing ion transport regulator 2, FXYD domain-containing ion transport regulator 2, HOMG2, hypomagnesemia 2, renal, MGC12372, Na(+)/K(+) ATPase subunit gamma, Sodium pump gamma chain, sodium/potassium-transporting ATPase subunit gamma, Sodium-potassium-ATPase, gamma polypeptide | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction