missing translation for 'onlineSavingsMsg'
Learn More

UFD1L Monoclonal Antibody (OTI3F1), TrueMAB™, OriGene

Product Code. p-200015936
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17543623 30 μL 30µL
17523623 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17543623 Supplier Origene Technologies Supplier No. TA804191S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UFD1L Monoclonal antibody specifically detects UFD1L in Human samples. It is validated for Western Blot

Specifications

Antigen UFD1L
Applications Western Blot
Classification Monoclonal
Clone OTI3F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CETN1
Gene Accession No. Q12798
Gene Alias RGD:619822}; UB fusion protein 1; UB fusion protein 1 {ECO:0000250; ubiquitin fusion degradation 1 like; ubiquitin fusion degradation 1 like (yeast); Ubiquitin fusion degradation protein 1; Ubiquitin fusion degradation protein 1 homolog; ubiquitin fusion degradation protein 1 homolog {ECO:0000250; Ubiquitin recognition factor in ER-associated degradation protein 1; UFD1; UFD1L; ufd1l {ECO:0000312; UniProtKB:Q92890}
Host Species Mouse
Immunogen Full length human recombinant protein of human CETN1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1068
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.