missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UFL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585467

Product Code. 16342405

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (92%), Rat (93%), Pig (96%), Chicken (80%), Rhesus Monkey (99%), Bovine (96%). Positive Control: 293T, A431, HeLa, HepG2, Jurkat, Raji, NCI-H929. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

UFL1 (UFM1 Specific Ligase 1) is a Protein Coding gene. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. Gene Ontology (GO) annotations related to this gene include ligase activity and UFM1 transferase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UFL1
Polyclonal
Unconjugated
Ufl1
1810074P20Rik; AI429228; E3 UFM1-protein ligase 1; E3 UFM1-protein transferase 1; Kiaa0776; LZAP-binding protein; Maxer; mKIAA0776; Multiple alpha-helix protein located at ER; NLBP; Novel LZAP-binding protein; RCAD; Regulator of C53/LZAP and DDRGK1; Regulator of CDK5RAP3 and DDRGK1; RP3-393D12.1; UFL1; UFM1 specific ligase 1; UFM1-specific ligase 1
Rabbit
Antigen affinity chromatography
RUO
23376
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
O94874
Ufl1
Recombinant protein encompassing a sequence within the N-terminus region of human KIAA0776. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.