missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGCGL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13420-1-AP-20UL
This item is not returnable.
View return policy
Description
UDP-glucose:glycoprotein glucosyltransferase (UGT) is a soluble protein of the endoplasmic reticulum (ER) that selectively reglucosylates unfolded glycoproteins, thus providing quality control for protein transport out of the ER.Specifications
| UGCGL2 | |
| Polyclonal | |
| Unconjugated | |
| UGGT2 | |
| HUGT2, UGCGL2, UGGT2, UGT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55757 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NYU1 | |
| UGGT2 | |
| UGCGL2 Fusion Protein Ag4189 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction