missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGDH Monoclonal Antibody (OTI2C11), TrueMAB™, OriGene
Description
The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluro, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia.
Specifications
Specifications
| Antigen | UGDH |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | UGDH |
| Gene Accession No. | O60701 |
| Gene Alias | GDH, UDP-GlcDH, UDPGDH, UGD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?