Learn More
UGGT1 Polyclonal Antibody
GREENER_CHOICE
UGGT1 Polyclonal Antibody
Brand:  Invitrogen PA5103025
Product Code. 16699474
  • € 502.00 / 100µL
Description
UGGT1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UGGT1
Polyclonal
Unconjugated
Uggt1
0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr
Rabbit
Affinity Chromatography
RUO
56886
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NYU2
Uggt1
A synthesized peptide derived from human UGGT1(Accession Q9NYU2), corresponding to amino acid residues E160-F210.