missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UGRP1 Monoclonal Antibody (K49035_3H11), Invitrogen™

Product Code. 30202097
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30202097 50 μL 50µg
30197842 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30202097 Supplier Invitrogen™ Supplier No. MA555673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: FLINKVPLPV DKLAPLPLDN ILPFMDPLKL LLKTLGISVE HLVEGLRKCV NELGPEASEA VKKLLEALSH LV

UGRP1 Monoclonal antibody specifically detects UGRP1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGRP1
Applications Western Blot
Classification Monoclonal
Clone K49035_3H11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SCGB3A2
Gene Accession No. Q920H1, Q96PL1
Gene Alias LU103; Pneumo secretory protein 1; PNSP1; PnSP-1; SCGB3A2; Secretoglobin family 3A member 2; secretoglobin, family 3A, member 2; UGRP1; UGRP1pnSP-1; UNQ566/PRO1128; Uteroglobin-related protein 1
Gene Symbols SCGB3A2
Host Species Mouse
Immunogen Recombinant human SCGB3A2 (immunogen range is 22-93).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117156, 117158
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.