missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UGT2B Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142710

Product Code. 17868322

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Additional Information: This antibody is NOT expected to cross-react with other Ugt2 proteins. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

UDPGT is of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. 2B2 acts on various endogenous steroids, especially etiocholanolone and androsterone.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UGT2B
Polyclonal
Unconjugated
Ugt2b
Ugt2b
Peptide with sequence C-KLDFLSALEEVIDN.
100 μg
Primary
Rat
Antibody
IgG
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P08541
Goat
Ammonium sulfate precipitation
RUO
24862
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.