missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94881-0.02ml
This item is not returnable.
View return policy
Description
UGT2B4 Polyclonal antibody specifically detects UGT2B4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UGT2B4 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| family 2, beta-4, UDP glucuronosyltransferase 2 family, polypeptide B4, UDP glycosyltransferase 2 family, polypeptide B4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 280-480 of human UGT2B4 (NP_066962.2). LHCKPAKPLPKEMEEFVQSSGENGVVVFSLGSMVSNTSEERANVIASALAKIPQKVLWRFDGNKPDTLGLNTRLYKWIPQNDLLGHPKTRAFITHGGANGIYEAIYHGIPMVGVPLFADQPDNIAHMKAKGAAVSLDFHTMSSTDLLNALKTVINDPLYKENAMKLSRIHHDQPVKPLDRAVFWIEFVMRHKGAKHLRVAA | |
| 0.02 mL | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 7363 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction