missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | UGT2B4 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287840
|
Novus Biologicals
NBP2-54718 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653899
|
Novus Biologicals
NBP2-54718-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UGT2B4 Polyclonal specifically detects UGT2B4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| UGT2B4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 7363 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:FVGGLHCKPAKPLPKEMEEFVQSSG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| family 2, beta-4, UDP glucuronosyltransferase 2 family, polypeptide B4, UDP glycosyltransferase 2 family, polypeptide B4 | |
| UGT2B4 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title