missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B4, Mouse anti-Human, Clone: 2H6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007363-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: KVLVWPTEFSHWMNIKTILDELVQRGHEVTVLASSASISFDPNSPSTLKFEVYPVSLTKTEFEDIIKQLVKRWAELPKDTFWSYFSQVQEIMWTFNDILRSpecifications
| UGT2B4 | |
| Monoclonal | |
| Unconjugated | |
| UDP glucuronosyltransferase 2 family, polypeptide B4 | |
| UGT2B11 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2H6 | |
| In 1x PBS, pH 7.4 | |
| NM_021139 | |
| UGT2B4 | |
| UGT2B4 (NP_066962.1, 25 a.a. ∼ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7363 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction