missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15155
This item is not returnable.
View return policy
Description
UGT2B7 Polyclonal specifically detects UGT2B7 in Human samples. It is validated for Western Blot.
Specifications
| UGT2B7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| UGT2B7 | |
| Recombinant protein encompassing a sequence within the center region of human UGT2B7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 7364 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| UDP glucuronosyltransferase 2 family, polypeptide B73,4-catechol estrogen specific, UDP glycosyltransferase 2 family, polypeptide B7, UGT2B9family 2, beta-7, UGTB2B9 | |
| Rabbit | |
| 61 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction