missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT3A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06222-100ul
This item is not returnable.
View return policy
Description
UGT3A1 Polyclonal antibody specifically detects UGT3A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UGT3A1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 2.4.1, EC 2.4.1.17, FLJ26528, FLJ34658, UDP glycosyltransferase 3 family, polypeptide A1, UDP-glucuronosyltransferase 3A1, UDPGT 3A1 | |
| Produced in rabbits immunized with E. coli-derived Human UGT3A1 fragment. | |
| 100 μg | |
| Lipid and Metabolism | |
| 133688 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction