missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASA1 Monoclonal Antibody (OTI2H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505900
This item is not returnable.
View return policy
Description
GTPase Activating Protein (GAP) is capable of stimulating the hydrolysis of GTP by ras p21 proteins, though GAP has little effect on the oncogenic forms of ras. It is also known that several tyrosine kinases such as platelet derived growth factor receptor and epidermal growth factor receptor are involved in the tyrosine phosphorylation of GAP. It has therefore been suggested that GAP may provide a connection or link between growth factor receptors and the ras p21 family.Specifications
| RASA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20936 | |
| RASA1 | |
| Full length human recombit protein of human RASA1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2H1 | |
| Unconjugated | |
| RASA1 | |
| CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5921 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction