missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UIP1 Antibody (OTI1E8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74782B
This item is not returnable.
View return policy
Description
UIP1 Monoclonal specifically detects UIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| UIP1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| HAUS7 | |
| Full length recombinant protein of human UIP1 (NP_059988) produced in HEK293T cells. | |
| 0.1 mL | |
| 55559.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 26S proteasome-associated UCH37-interacting protein 1, expressed-Xq28STS protein, HAUS augmin-like complex subunit 7, HAUS augmin-like complex, subunit 7, UCH37 interacting protein 1, UCHL5 interacting protein, UCHL5-interacting protein, UCHL5IPHSXQ28ORF, UIP126S proteasome-associated UCH interacting protein 1, X-linked protein STS1769 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction