missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-2 Antibody (MM0593-7F33), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11987JF646
This item is not returnable.
View return policy
Description
ULBP-2 Monoclonal antibody specifically detects ULBP-2 in Human samples. It is validated for Flow Cytometry
Specifications
| ULBP-2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2 | |
| Human recombinant ULBP-2 | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 80328 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry | |
| MM0593-7F33 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction