missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-3 Antibody (166510) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1517B
This item is not returnable.
View return policy
Description
ULBP-3 Monoclonal antibody specifically detects ULBP-3 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| ULBP-3 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 166510 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| NKG2D ligand 3, RAET1N, UL16 binding protein 3 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-3 | |
| 0.1 mL | |
| Immunology | |
| 79465 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction