missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-3 Antibody (MM0594-6E12), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11988R
This item is not returnable.
View return policy
Description
ULBP-3 Monoclonal specifically detects ULBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ULBP-3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ULBP3 | |
| Human recombinant ULBP-3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0594-6E12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NKG2D ligand 3, RAET1N, UL16 binding protein 3 | |
| Mouse | |
| Protein G purified | |
| Immunology | |
| 79465 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction