Learn More
ULBP-4/RAET1E Antibody (RAET1E 50/3), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50267APCCY7
Code produit. 18197798
  • € 459.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
ULBP-4/RAET1E
Monoclonal
APC-Cyanine7
PBS with 0.05% Sodium Azide
RAET1E
Recombinant His-tagged protein to human extra-cellular domain of ULBP-4/RAET1E, grown in E.coli
0.1 mL
Primary
Human
IgG2b
Flow Cytometry
RAET1E 50/3
Flow Cytometry
bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308
Mouse
Protein A purified
Immunology
135250
Store at 4°C in the dark. Do not freeze.