missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ULK3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142822
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
Specifications
| ULK3 | |
| Polyclonal | |
| Unconjugated | |
| ULK3 | |
| 1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 25989 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6PHR2 | |
| ULK3 | |
| Peptide with sequence C-KMRESRWEADTLDK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction