Learn More
Invitrogen™ ULK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110548
Description
Immunogen sequence: MENFILYEEI GRGSKTVVYK GRRKGTINFV AILCTDKCKR PEITNWVRLT REIKHKNIVT FHEWYETSNH LWLVVELCTG GSLKTVIAQD ENLPEDVVRE FGIDLISGLH HLHKLGILFC DISPRKILLE GPGTLKFSNF CLAKVEGENL EEFFALVAAE EGGGDNGENV LKKSMKSRVK GSPVYTAPEV VRGADFSISS DLWSLGCLLY EMFSGKPPFF SESISELTEK ILCEDPLPPI PKDSSRPKAS SDFINLLDGL LQRDPQKRLT WTRLLQHSFW KKAFAGADQE.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. ULK4 (unc-51-like kinase 4) is a 1,275 amino acid protein that contains one protein kinase domain and one HEAT repeat and belongs to the Ser/Thr protein kinase family. Although containing what is thought to be a catalytically inactive domain, ULK4 may play a role in the ATP-dependent phosphorylation of target proteins.
Specifications
| ULK4 | |
| Polyclonal | |
| Unconjugated | |
| ULK4 | |
| 4932415A06Rik; A730098P15; FAM7C1; REC01035; RGD1565726; serine/threonine-protein kinase ULK4; ULK4; unc-51 like kinase 4; unc-51-like kinase 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 209012, 363167, 54986 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3V129, Q96C45 | |
| ULK4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human ULK4 (NP_060356.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.