Learn More
Ultra Low Sulfur Diesel Fuel Blank, VHG™
Description
Ultra Low Sulfur Diesel Fuel Blank
Specifications
Specifications
| Chemical Name or Material | Blank |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Blank |
| Shipping Condition | Room Temperature |
| IUPAC Name | Blank |
| Solvent or Matrix | #2 Diesel Fuel |
| Quantity | 0.5 gal |