missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra-Pure, Chem-Lab

Catalog Number p-8014514
Click to view available options
Quantity:
5 L
10 L
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Ultra purified for laboratory use, AAS and ICP. Water, Ultra-Pure, Chem-Lab meets ASTM D-94 and D-129 standards. It is UV filtrated with LF < 1μS – 0.4μm. Available in a range of quantities: 2.5, 5 and 10L.

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.016g/mol
Grade Ultra Pure
Vapor Pressure 17.5mm Hg at 20°C
Filtered Through LF < 1μS – o0.4μm, UV filtrated
Density 1.00g/mL
Quantity 5 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'