missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ UltraScence Pico Plus Western Substrate

Product Code. 30162602
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30162602 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30162602 Supplier R&D Systems™ Supplier No. NBP333176
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Luminol-based enhanced chemiluminescent substrate

The UltraScence Pico Plus Western Substrate, as a luminol-based enhanced chemiluminescent substrate, is sensitive and compatible with conducting immunoblots with horseradish peroxidase (HRP) - conjugated secondary antibodies. The low picogram or high femtogram detection of antigen is enabled by UltraScence Pico Plus Western Substrate's excellent sensitivity and long signal duration. Further, its long chemiluminescent signal duration makes both digital and film-based imaging possible without any loss of the signal. Appropriate primary and secondary antibody dilutions are suggested for attaining optimal signal intensity and duration.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Product Line Novus
Quantity 100 mL
Content And Storage Store at 4°C. Do not freeze.
Target UltraScence Pico Plus Western Substrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.