Learn More
Bandelin Electronic™ Stammopur™ 24 Ultrasonic Cleaning Fluid
Click to view available options
Volume (Metric):
2 L
Unit Size:
2 L
Description
- Active against bacteria (including Tb.-B.) and fungi
- Virucidal against Vaccinia, BVDV, H5N1, HBV, HCV, HIV
- Mildly alkaline, pH 9.4 (1%)
- Dosage 1-2%
- Cleans and disinfects items such as technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
| pH | 9.4 |
| Certifications/Compliance | VAH |
| Concentration | 1%/15min. |
| Container Type | Bottle |
| Volume (Metric) | 2 L |
Safety and Handling
Product Identifier
- STAMMOPUR 24
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Butan-1, 4-diol, lactic acid, piperazine, alkylpolyglycoside