missing translation for 'onlineSavingsMsg'
Learn More

Umbralisib R-enantiomer, MedChemExpress

Product Code. 16450156
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
This item is not returnable. View return policy

Product Code. 16450156

Brand: Medchem Express HY12279F

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Umbralisib R-enantiomer (TGR-1202 R-enantiomer) is a PI3Kδ inhibitor, which is the less active enantiomer of TGR-1202.

Umbralisib R-enantiomer (TGR-1202 R-enantiomer) is a PI3Kδ inhibitor, which is the less active enantiomer of TGR-1202.

Specifications

Chemical Name or Material Umbralisib R-enantiomer
CAS 1532533-69-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H24F3N5O3
Quantity 2 mg
Synonym TGR-1202 R-enantiomer RP5264 R-enantiomer
SMILES O=C1C(C2=CC=CC(F)=C2)=C([C@@H](C)N3N=C(C4=CC=C(OC(C)C)C(F)=C4)C5=C(N)N=CN=C53)OC6=CC=C(F)C=C16
Molecular Weight (g/mol) 571.55
Formula Weight 571.55
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.