missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC119B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524504
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Xenopus and zebrafish based on sequence homology.
Mouse Unc119b is related closely to Unc119, a homolog of C. elegans unc119 protein which was first discovered in C. elegans on the basis of a spontaneous mutation affecting locomotion, feeding behavior and chemosensation. Mouse Unc119b shares 90% sequence identity with human UNC119b protein. UNC119b is a myristoyl-binding protein that acts as a cargo adapter: specifically binds the myristoyl moiety of a subset of N-terminally myristoylated proteins and is required for their localization. Myristoyl-binding activity of UNC119b is required to localize NPHP3 to the primary cilium.
Specifications
| UNC119B | |
| Polyclonal | |
| Unconjugated | |
| UNC119B | |
| AA407659; AI414892; POC7 centriolar protein homolog B; POC7B; Protein unc-119 homolog B; protein unc-119 homolog B; LOW QUALITY PROTEIN: protein unc-119 homolog B; unc119 homolog B; unc-119 homolog B; unc-119 homolog B (C. elegans); unc-119 lipid binding chaperone B; UNC119B | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 106840, 84747 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NIH7, Q8C4B4 | |
| UNC119B | |
| KLH conjugated synthetic peptide between 207-236 amino acids from the C-terminal region of human U119B. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction