missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC13B Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142610
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_074053.1; NP_001036044.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. Is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-depending refilling of readily releasable vesicle pool (RRP). Essential for synaptic vesicle maturation in a subset of excitatory/glutamatergic but not inhibitory/GABA-mediated synapses.
Specifications
| UNC13B | |
| Polyclonal | |
| Unconjugated | |
| UNC13B | |
| homolog of rat Munc13 (diacylglycerol-binding); munc13; Munc13-1; Munc13-2; protein unc-13 homolog B; UNC13; unc13 homolog 1; UNC-13 homolog 1; unc-13 homolog A; unc-13 homolog B; unc-13 homolog B (C. elegans); Unc13a; Unc13b; Unc13h1; Unc13h2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10497 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O14795 | |
| UNC13B | |
| Peptide with sequence PLRDVAAKGSC. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction