missing translation for 'onlineSavingsMsg'
Learn More

UNC13D/Munc 13-4 Antibody, Novus Biologicals™

Product Code. 18631098 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18631098 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18631098 Supplier Novus Biologicals Supplier No. NBP25782725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UNC13D/Munc 13-4 Polyclonal specifically detects UNC13D/Munc 13-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC13D/Munc 13-4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans)
Gene Symbols UNC13D
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SFHLDMWDLDTVESVRQKLGELTDLHGLRRIFKEARKDKGQDDFLGNVVLRLQDLRCREDQWYPLEPRTETYPDRGQCHLQFQLIHKRRATSASRSQPSYTVHLHLLQQLVSHEVTQHEAGSTSWDGSLSPQAATVLFLHATQKDLSD
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201294
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.