missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC45B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21640-1-AP-150UL
This item is not returnable.
View return policy
Description
UNC45B plays a role in myoblast fusion and sarcomere organization.Specifications
| UNC45B | |
| Polyclonal | |
| Unconjugated | |
| UNC45B | |
| CMYA4, Protein unc 45 homolog B, SMUNC45, unc 45 homolog B (C. elegans), Unc 45B, UNC45, UNC45B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146862, 217012, 303373 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CGY6, Q8IWX7 | |
| Unc45b | |
| UNC45B Fusion Protein Ag16294 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction