missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC50 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93822-0.02ml
This item is not returnable.
View return policy
Description
UNC50 Polyclonal antibody specifically detects UNC50 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UNC50 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| geal-6 membrane-associated high-copy suppressor 1, GMH1, hGMH1, PDLs22, Periodontal ligament-specific protein 22, Protein GMH1 homolog, protein unc-50 homolog, unc-50 homolog (C. elegans), unc-50 related, UNCLDKFZp564G0222, Uncoordinated-like protein, URP | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UNC50 (NP_054763.2). MLPSTSVNSLVQGNGVLNSRDAARHTAGAKRYKYLRRLFRFRQMDFEFAAWQMLYLFTSPQRVYRNFHYRKQTKDQWARDDPAFLVLLSIWLCVSTIGFG | |
| 0.02 mL | |
| Neuroscience | |
| 25972 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction