missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UNC5A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15990135
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15990135 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15990135 Supplier Invitrogen™ Supplier No. PA567592

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

The UNC5H family of proteins act as transmembrane receptors for netrin-1 and play a crucial role in axon guidance and migration of neural cells. Additionally, when cleaved by a caspase to produce an intracellular fragment containing a death domain, UNC5H receptors induce apoptosis. This activity is blocked by the binding of netrin-1. In the absence of netrin-1, UNC5H receptors act as tumor suppressors by inhibiting anchorage-independent growth and invasion, but mutation of these receptors provides a potential mechanism for tumorigenicity. The expression of UNC5H receptors is downregulated in multiple carcinomas, including colorectal, breast, ovary, uterus, stomach, lung and kidney cancers. UNC5H1, also known as UNC5HA (unc-5 homolog A), is a member of the UNC5H family of proteins and is localized to the cell membrane. Three isoforms of UNC5H1 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Unc5a
Gene Accession No. O08721, Q6ZN44, Q8K1S4
Gene Alias Kiaa1976; mKIAA1976; netrin receptor UNC5A; netrin receptor Unc5h1; protein unc-5 homolog 1; Protein unc-5 homolog A; transmembrane receptor Unc5H1; unc5 (C.elegans homolog) a; unc5 homolog 1; unc-5 homolog 1; unc-5 homolog A; unc-5 homolog A (C. elegans); unc-5 netrin receptor A; UNC5A; Unc5h1; UNC5H1FLJ16449
Gene Symbols Unc5a
Host Species Rabbit
Immunogen Synthesized peptide derived from human UNC5A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107448, 60629, 90249
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.