Learn More
Invitrogen™ UNC5CL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582536
Description
Immunogen sequence: MHTFQDGLET KYMEILRFQA SEEESWAAPP PVSQPPPCNR LPPELFEQLR MLLEPNSITG NDWRRLASHL GLCGMKIRFL SCQRSPAAAI LELFEEQNGS LQELHYLMTV MERLDCASAI QNYLSGTHGG SPGPERGGAR DNQGLELDEK Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 87%.
UNC5CL is a membrane protein belonging to theUNC-5 family with a death domain and a ZU5 domain and is known to interact with p65/RELA and NFKB1. UNC5CL is known to interact with subunit p105 of NF-kappaB and transactivator p65 and is thus involved in inhibition of NF-kappaB activation and NF-kappaB-dependent transcription by inhibiting the binding of NF-kappaB to its target sequence (providing an alternative regulatory mechanism for NF-kappaB-mediated transcription). This regulatory protein sensitizes cells to apoptosis induced by TNF and TRAIL.
Specifications
| UNC5CL | |
| Polyclonal | |
| Unconjugated | |
| UNC5CL | |
| 2510009H09Rik; BB155270; MGC34763; MUXA; OTTHUMP00000016362; protein unc-5 homolog C-like; unc-5 family C-terminal like; unc-5 homolog C (C. elegans)-like; UNC5CL; UNC5C-like protein; zu-5; ZU5 and death domain containing; ZU5 and death domain-containing inhibitor of NF-kB; ZU5 and death domain-containing protein; ZUD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IV45 | |
| UNC5CL | |
| Recombinant protein corresponding to Human UNC5CL. Recombinant protein control fragment (Product #RP-90179). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.