missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC79 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584881
This item is not returnable.
View return policy
Description
Immunogen sequence: SFALPEMSLD DHPDPGTEGE KPGELMPSSG AKTVLLKVPE DAENPTESEK PDTSAESDTE QNPERKVEED GAEESEFKIQ IVPRQRKQRK IAVSA Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 80%.
The NALCN channel is responsible for Na(+) leak currents. The protein encoded by this gene, along with UNC80, is an accessory subunit of the NALCN channel that contributes to the Ca(2+) sensitivity of the channel. [provided by RefSeq, Sep 2016].
Specifications
| UNC79 | |
| Polyclonal | |
| Unconjugated | |
| UNC79 | |
| KIAA1409; protein unc-79 homolog; UNC79; unc-79 homolog (C. elegans) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57578 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2D8 | |
| UNC79 | |
| Recombinant protein corresponding to Human UNC79. Recombinant protein control fragment (Product #RP-108591). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction