missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC93A Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81858UV
This item is not returnable.
View return policy
Description
UNC93A Polyclonal antibody specifically detects UNC93A in Human samples. It is validated for Western Blot, ELISA
Specifications
| UNC93A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54346 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 350 | |
| dJ366N23.1, dJ366N23.1 (putative C. elegans UNC-93 (protein 1, C46F11.1) LIKE), dJ366N23.2, dJ366N23.2 (putative C. elegans UNC-93 (protein 1, C46F11.1) C-terminal LIKE), unc93 homolog A, unc-93 homolog A (C. elegans) | |
| UNC93A antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human UNC93A. The immunogen is located within the last 50 amino acids of UNC93A. Amino Acid Squence: CVESKNPIRPHAPGQV | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction