Learn More
Köttermann Underbench Acid and Base Cabinet
Product Code. 17312179
Change view
Click to view available options
Dimensions (D x W x H):
515 x 600 x 720 mm
515 x 900 x 720 mm
520 x 450 x 720 mm
515 x 1100 x 720 mm
515 x 1200 x 720 mm
515 x 1400 x 720 mm
Configuration:
1 Left-hinged Door, 1 Drawer for Containers
1 Left-hinged Door, 1 Pull-out
1 Left-hinged Door, 2 Drawers
1 Left-hinged Door, 2 Pull-outs
1 Right-hinged Door, 1 Drawer for Containers
1 Right-hinged Door, 1 Pull-out
1 Right-hinged Door, 2 Drawers
1 Right-hinged Door, 2 Pull-outs
2 Doors, 3 Drawers
2 Doors, 3 Drawers (1 for Containers)
2 Doors, 4 Drawers
Includes:
No Ventilation Fan
Ventilation Fan
Unit Size:
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Configuration Includes unitSize
17312179 520 x 450 x 720 mm 1 Left-hinged Door, 1 Pull-out No Ventilation Fan Each
17392169 520 x 450 x 720 mm 1 Left-hinged Door, 2 Pull-outs No Ventilation Fan Each
17302179 520 x 450 x 720 mm 1 Right-hinged Door, 2 Pull-outs No Ventilation Fan Each
17322179 520 x 450 x 720 mm 1 Right-hinged Door, 1 Pull-out No Ventilation Fan Each
17332169 515 x 600 x 720 mm 1 Left-hinged Door, 1 Drawer for Containers No Ventilation Fan Each
17332159 515 x 600 x 720 mm 1 Left-hinged Door, 2 Drawers No Ventilation Fan Each
17342169 515 x 600 x 720 mm 1 Right-hinged Door, 1 Drawer for Containers No Ventilation Fan Each
17342159 515 x 600 x 720 mm 1 Right-hinged Door, 2 Drawers No Ventilation Fan Each
17392159 515 x 900 x 720 mm 2 Doors, 3 Drawers Ventilation Fan Each
17352169 515 x 900 x 720 mm 2 Doors, 3 Drawers (1 for Containers) No Ventilation Fan Each
17352159 515 x 900 x 720 mm 2 Doors, 4 Drawers No Ventilation Fan Each
17302169 515 x 1100 x 720 mm 2 Doors, 3 Drawers Ventilation Fan Each
17362169 515 x 1100 x 720 mm 2 Doors, 3 Drawers (1 for Containers) No Ventilation Fan Each
17362159 515 x 1100 x 720 mm 2 Doors, 4 Drawers No Ventilation Fan Each
17312169 515 x 1200 x 720 mm 2 Doors, 3 Drawers Ventilation Fan Each
17372169 515 x 1200 x 720 mm 2 Doors, 3 Drawers (1 for Containers) No Ventilation Fan Each
17372159 515 x 1200 x 720 mm 2 Doors, 4 Drawers No Ventilation Fan Each
17322169 515 x 1400 x 720 mm 2 Doors, 3 Drawers Ventilation Fan Each
17382169 515 x 1400 x 720 mm 2 Doors, 3 Drawers (1 for Containers) No Ventilation Fan Each
17382159 515 x 1400 x 720 mm 2 Doors, 4 Drawers No Ventilation Fan Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 17312179 Supplier Köttermann Supplier No. 3770023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Constructed with durable polypropylene, this Underbench Acid and Base Cabinet is built to endure the aggressive conditions produced by corrosive acid fumes. Liquid-tight pull-out trays offer easy access while minimizing health and safety risks.
  • Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
  • Cabinet carcasses and pull-out trays made of resistant polypropylene
  • Liquid-tight drawers on low-friction PTFE strips with pull-out stop
  • Removal of aggressive vapors via optional ventilation with an external fan
  • Corrosion-resistant locks and hinges
  • Ideal for installation under high-performance fume cupboards
Specifications
Configuration 1 Left-hinged Door, 1 Pull-out
Dimensions (D x W x H) 520 x 450 x 720 mm
Door Style Hinged, Drawer
For Use With (Application) Acid and Base Storage
Includes No Ventilation Fan
Color White
Material Polypropylene